Learn More
Memmert™ Entry Port 23 mm, Standard Position Left Side Centre/Top, For S75
Use these entry ports with convection ovens. Memmert™ Convection and Universal Oven Entry Points can be closed by flap in special positions at the back.
Brand: Memmert™ S75F1
Additional Details : Weight : 0.01100kg
Description
- Entry port with clear diameter
- For Memmert Ovens
Specifications
Models 30, 55, 75, 110, 160, 260, 450, 750, 1060 | |
23 mm | |
Entry Port | |
Stainless Steel |