Learn More
Memmert™ Entry Port 57 mm for UN450plus
Optional entry port for Memmert Ovens.
Brand: Memmert™ UN450plusF8
Additional Details : Weight : 0.01200kg
Description
- Entry port with 57 mm diameter
- Flap closure
- Custom position TBC in the back
Specifications
UN450plus Oven | |
Entry Port | |
57 mm |