Learn More
Memmert™ Entry Port Right Centre/Centre
For introducing connections at the side and can be closed by flap
138.00€ - 240.00€
Specifications
Color Clear
Diameter (Metric) 23 mm
View More Specs
Products 10
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
17110275
View Documents
Memmert™
IPP750ecoF2
IPP750eco
138.00€
Each
Estimated Shipment: 12-08-2025
Add to basket
17129834
View Documents
Memmert™
HPP410ecoF2
HPP410eco
240.00€
Each
Estimated Shipment: 12-08-2025
Add to basket
17139694
View Documents
Memmert™
HPP110ecoF2
HPP110eco
240.00€
Each
Estimated Shipment: 02-07-2025
Add to basket
17150065
View Documents
Memmert™
IPP1060ecoF2
IPP1060eco
138.00€
Each
Estimated Shipment: 12-08-2025
Add to basket
17160105
View Documents
Memmert™
IPP110ecoF2
IPP110eco
138.00€
Each
Estimated Shipment: 02-07-2025
Add to basket
17179644
View Documents
Memmert™
HPP1060ecoF2
HPP1060eco
240.00€
Each
Estimated Shipment: 12-08-2025
Add to basket
17179784
View Documents
Memmert™
HPP260ecoF2
HPP260eco
240.00€
Each
Estimated Shipment: 12-08-2025
Add to basket
17189874
View Documents
Memmert™
HPP750ecoF2
HPP750eco
240.00€
Each
Estimated Shipment: 12-08-2025
Add to basket
17190185
View Documents
Memmert™
IPP260ecoF2
IPP260eco
138.00€
Each
Estimated Shipment: 12-08-2025
Add to basket
17190225
View Documents
Memmert™
IPP410ecoF2
IPP410eco
138.00€
Each
Estimated Shipment: 12-08-2025
Add to basket
Description
Description
  • Standard position right centre/centre
  • Not for model sizes 110 and 260 with light module
  • Not for model size 110 with light module
Specifications
Specifications
Clear
23 mm
Documents
Product Certifications
Product Content Correction
Product Title
Memmert™ Entry Port Right Centre/Centre