missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Enzymatic Digest Casein Hydrolysate, Thermo Scientific Chemicals
Enzymatic digest of casein.
Brand: Thermo Scientific J12855.P5
This item is not returnable.
View return policy
Description
- Synonym: Peptones
- pH: 6.6 to 7.5
- CAS: 65072-00-6
- MDLNumber: MFCD00282824
Specifications
Casein hydrolysate | |
2.2 kg (5 lb.) | |
Powder |
6.6 to 7.5 | |
Plastic bottle |
RUO – Research Use Only