Learn More
BRAND™ EPDM Rubber Conical Gaskets
Place between filter funnel and filter flask for a secure fit.
Brand:  BRAND™ 462932
100.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10049900
  • 103.00€ / Pack of 10
In Stock
Description
  • Rubber (EPDM) construction
  • Conical shape
  • Provides a secure fit between filter funnel and filter flask
  • Supplied in packs of 10
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Used for Filter Funnels and Filter Flasks
10
EPDM
50 mm
32 mm
Rubber Conical Gasket
Product Title
BRAND™ EPDM Rubber Conical Gaskets > Diameter: 50mm