Learn More
BRAND™ EPDM Rubber Conical Gaskets
Place between filter funnel and filter flask for a secure fit.
Brand: BRAND™ 462723
82.50 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Rubber (EPDM) construction
- Conical shape
- Provides a secure fit between filter funnel and filter flask
- Supplied in packs of 10
Specifications
Used for Filter Funnels and Filter Flasks | |
10 | |
EPDM |
35 mm | |
20 mm | |
Rubber Conical Gasket |
Product Content Correction
Product Title
BRAND™ EPDM Rubber Conical Gaskets > Diameter: 35mm