Learn More
Lauda™ Hose
Material: EPDM
Brand:  Lauda™ RKJ 112
Product Code. 10729474
  • 28.50€ / 1m
Estimated Shipment: 20-01-2025
Description
  • For heat transfer of all liquids
Specifications
Specifications
10°C to 90°C
EPDM Rubber
12 mm
Hose
16 mm