Learn More
Eppendorf™ epMotion™ Reservoir, 195 mL
Low-dead-volume, PCR-clean reservoirs are constructed of polypropylene and feature a single well with 96 cavities.
Brand: Eppendorf™ 0030126556
This item is not returnable.
View return policy
Description
- Holds up to 195 mL reagent
- Tapered shape for full reagent recovery
- Available as tempered version (cooling or warming)
- Modules to increase tip space
Specifications
195 mL | |
Non-sterile | |
4 x 5 reservoirs | |
96 cavities |
Polypropylene | |
Low Profile Reservoir | |
96 |