Learn More
Eppendorf™ epMotion™ Reservoir, 195 mL
Low-dead-volume, PCR-clean reservoirs are constructed of polypropylene and feature a single well with 96 cavities.
Brand:  Eppendorf™ 0030126556
Product Code. 30040080
  • 389.00€ / Pack of 20
In Stock
Description
  • Holds up to 195 mL reagent
  • Tapered shape for full reagent recovery
  • Available as tempered version (cooling or warming)
  • Modules to increase tip space
TRUSTED_SUSTAINABILITY
Specifications
Specifications
195 mL
Non-sterile
4 x 5 reservoirs
96 cavities
Polypropylene
Low Profile Reservoir
96