Learn More
Eppendorf™ Lid for ThermoMixer™ F1.5 and FP Product Code.: 13508480
In Stock
to see stock.
Add to basket
Eppendorf™ Lid for ThermoMixer™ F1.5 and FP
Product Code. 13508480 Shop All Eppendorf Products
0.5, 1.5, 2mL SmartBlocks, 96 and 384-well PCR plates
Click to view available options
For Use With (Equipment):
0.5, 1.5, 2mL SmartBlocks, 96 and 384-well PCR plates
Product Code. 13508480
Brand: Eppendorf™ 5363000233
In Stock
to see stock.
Edge
Add to basket
Add to basket
Use this lid with ThermoMixer™ F1.5 and SmartBlocks™ 0.5mL, 1.5mL, 2.0mL, plates PCR 96, PCR 384. Eppendorf™ Lid for ThermoMixer™ F1.5 and FP is exchangeable in seconds.
Maintain accurate, uniform temperatures with individually sensor-controlled SmartBlocks.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) 0.5, 1.5, 2mL SmartBlocks, 96 and 384-well PCR plates
Type Accessory, Lid
Product Content Correction
Product Title
Eppendorf™ Lid for ThermoMixer™ F1.5 and FP >