Learn More
Thermo Scientific™ Crimper and Decrimper Jaw Sets for Programmable Electronic High-Power Crimp Stations
Optimize your Thermo Scientific™ Programmable Electronic High-Power Crimp Stations with these crimper and decrimper sets.
Brand:  Thermo Scientific™ ECMJ-20
344.66 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15287308
  • 346.38€ / Each
Description
Optimize your Thermo Scientific™ Programmable Electronic High-Power Crimp Station with these crimper and decrimper sets.
Available in a variety of sizes and formats.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 Ea.
Programmable Electronic High-Power Crimp Station, 20 mm Vial Closure
Industrial Chromatography
Electronic Crimper
20 mm
SDS
Documents Certificates
Product Title
Thermo Scientific™ Crimper and Decrimper Jaw Sets for Programmable Electronic High-Power Crimp Stations >