Learn More
ES/NO3 Nitrate Standard Solution, WTW™
Click to view available options
Quantity:
1000 mL
Specifications
Specifications
Concentration | 10g/L |
Quantity | 1000 mL |
Color | Colorless |
Boiling Point | 100°C |
Physical Form | Liquid |