Learn More
Notrax ESD Heel Grounder Product Code.: 17761918
Estimated Shipment: 30-05-2025
Add to basket
Notrax ESD Heel Grounder
Product Code. 17761918
Heel Grounder; For Use With: Connecting a Person with ESD Properties
Click to view available options
Heel Grounder; For Use With: Connecting a Person with ESD Properties
Product Code. 17761918
Brand: Notrax 055SHG0100
Estimated Shipment: 30-05-2025
Add to basket
Add to basket
Notrax ESD Heel Grounder is a device for connecting a walking or standing person to ground by using a conductive rubber thread to provide the necessary electrical contact between the person and the floor mat.
  • A device for connecting a walking or standing person to ground by using a conductive rubber thread to provide the necessary electrical contact between the person and the floor mat
  • Specialized line of ESD accessories to ensure a static free workplace and protect employees and sensitive electrical equipment
  • Use these accessories to connect table work surface, employees and conductive material to a common grounding point
  • This creates a balance between all items and personnel to ensure static electricity will flow to ground in a controlled manner
  • For use in ESD working zones to protect electrostatic sensitive components
Specifications
Product Type Heel Grounder
Material Wire
For Use With (Application) Connecting a Person with ESD Properties
Product Content Correction
Product Title
Notrax ESD Heel Grounder >