Learn More
Invitrogen™ Estrone-3-Glucuronide (E1G) Competitive ELISA Kit
Competitive ELISA
4635.00€
Specifications
Assay Sensitivity | 7.38 pg/mL |
---|---|
Description | Estrone-3-Glucuronide (E1G) Competitive ELISA Kit |
Conjugate | HRP |
Detection Method | Fluorescence |
Product Type | ELISA Kit |
Description
The Estrone-3 Glucuronide (E1G) ELISA quantitates estrone-3 glucuronide in extracted serum,plasma,urine,dried fecal extracts or cell culture medium.
Principle of the method
The Estrone-3-Sulfate (E1S) Competitive ELISA research-use-only kit uses a specifically generated antibody to measure Estrone-3-Sulfate (E1S) in a variety of matrices,including serum,plasma,urine,and fecal samples. The kit will quantitatively measure E1S present in diluted buffer samples and tissue culture media samples. An E1S standard is provided to generate a standard curve for the assay and all samples should be read off the standard curve. Standards or diluted samples are pipetted into a clear microtiter plate coated with an antibody to capture rabbit antibodies. An E1S-peroxidase conjugate is added to the standards and samples in the wells. The binding reaction is initiated by the addition of a polyclonal antibody to E1S to each well. After a 2-hour incubation the plate is washed and substrate is added. The substrate reacts with the bound E1S-peroxidase conjugate. After a short incubation,the reaction is stopped and the intensity of the generated color is detected in a microtiter plate reader capable of measuring at 450 nm.
Rigorous validation
Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity,specificity,precision,and lot-to-lot consistency. See manual for more information on validation.
Estrone-3-glucuronide (E1G) is the principle secreted form of circulating estradiol in mammals. Estrone-3-glucuronide (E1G) is identical across all species.Specifications
7.38 pg/mL | |
HRP | |
ELISA Kit | |
Human | |
Colorimetric Microplate Reader | |
3.8% | |
RUO | |
-20°C | |
1 hr. 20 min. |
Estrone-3-Glucuronide (E1G) Competitive ELISA Kit | |
Fluorescence | |
Fecal Extract, Serum, Supernatant, Urine | |
ELISA, Protein Assays, Aktivity Assays, Protein Biology, Reproductive Biology, Inflammation, Oxidative Stress, Metabolism, Cell Signaling, Epigenetics, Hematology, DNA Damage, Cancer Biology, Immunology, Neuroscience, Neurobiology, Steroid Hormone Detecti | |
5.6% | |
HRP | |
Reproductive Biology | |
Chemical | |
2 hr. 30 min. |