missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Estrone Competitive ELISA Kit

Competitive ELISA

4635.00€

Specifications

Assay Sensitivity 22.4 pg/mL
Description Estrone Competitive ELISA Kit
Conjugate HRP
Detection Method Fluorescence
Product Type ELISA Kit
View More Specs

Products 1
Product Code Brand Sufficient For Price Quantity & Availability  
Product Code Brand Sufficient For Price Quantity & Availability  
17835117
View Documents
Invitrogen™
EIA17E1X10
10 x 96 Assays
4635.00€
Pack of 10
Estimated Shipment: 31-01-2025
to see stock.
Add to basket
 
Description

Description

The Multispecies Estrone ELISA quantitates estrone in fecal extracts,urine,and tissue culture media.

Principle of the method
The Estrone Competitive ELISA research-use-only kit is designed to quantitatively measure estrone present in extracted dried fecal samples,urine,and tissue culture media samples. The kit is unique as it measures both non-conjugated estrone,estrone-3-sulfate,and estrone 3-glucuronide in urine and fecal samples with almost equal affinity,allowing for non-invasive testing of this steroid. An estrone standard is provided to generate a standard curve for the assay and all samples should be read off the standard curve. Standards or diluted samples are pipetted into a clear microtiter plate coated with an antibody to capture rabbit antibodies. An estrone-peroxidase conjugate is added to the standards and samples in the wells. The binding reaction is initiated by the addition of a polyclonal antibody to estrone to each well. After a 2-hour incubation the plate is washed and substrate is added. The substrate reacts with the bound estrone-peroxidase conjugate. After a short incubation,the reaction is stopped and the intensity of the generated color is detected in a microtiter plate reader capable of measuring at 450 nm.

Rigorous validation
Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity,specificity,precision,and lot-to-lot consistency. See manual for more information on validation.

Estrone is an aromatized C18 steroid with a 3-hydroxyl group and a 17-ketone, a major mammalian estrogen. it is converted from androstenedione directly, or from testosterone via estradiol. In humans, it is produced primarily by the cyclic ovaries, placenta, and the adipose tissue of men and postmenopausal women. Therapeutic estrone is the synthetic form of a naturally occurring estrogen estrone. Estrone diffuses through the cell membrane and binds to and subsequently activates the nuclear estrogen receptor found in the reproductive tract, breast, pituitary, hypothalamus, liver, and bone. The activated hormone-receptor complex gets translocated to the nucleus where it binds to the estrogen response element on the dna and promotes the transcription of target genes involved in the functioning of the female reproductive system and secondary sex characteristics. Estrone increases the hepatic synthesis of sex hormone binding globulin (shbg), thyroid-binding globulin (tbg), and other serum proteins, as well as suppresses the release of follicle-stimulating hormone (fsh) and luteinizing hormone (lh) from the anterior pituitary. [National Center for Biotechnology Information. PubChem Compound Database; CID=5870].
Specifications

Specifications

22.4 pg/mL
HRP
ELISA Kit
Human
Colorimetric Microplate Reader
4.7%
RUO
-20°C
1 hr. 20 min.
Estrone Competitive ELISA Kit
Fluorescence
Fecal Extract, Supernatant, Urine
ELISA, Protein Assays, Aktivity Assays, Protein Biology, Reproductive Biology, Inflammation, Oxidative Stress, Metabolism, Cell Signaling, Epigenetics, Hematology, DNA Damage, Cancer Biology, Immunology, Neuroscience, Neurobiology, Steroid Hormone Detecti
5.5%
HRP
Reproductive Biology
Human
2 hr. 30 min.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.