Learn More
Draeger Safety Ethanol Diffusion Tube
Draeger Safety Ethanol Diffusion Tube is a direct-reading diffusion tube used specifically for personal exposure monitoring.
Brand:  Draeger Safety 8101151
Product Code. 15613176
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Explore more special offers
Description
Description
  • Direct-reading diffusion tubes are used specifically for personal exposure monitoring
  • These tubes do not require the use of a gas detector pump because the contaminant molecules pass into the tube by diffusion processes and in this manner are brought into contact with the reagent systems
  • Exposure levels are easily read by the markings on the side of the tube
Specifications
Specifications
Ethanol 1000/a-D
Diffusion Tube
1 hr: 1000 to 25,000 ppm, 4 hr: 125 to 3100 ppm