missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1H-indazole-5-carboxylate, 95%, Thermo Scientific™

Product Code. 15405397
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 192944-51-7
Molecular Formula C10H10N2O2
Molecular Weight (g/mol) 190.20
MDL Number MFCD03426220
InChI Key SKABXDPLIJIWLR-UHFFFAOYSA-N
Synonym 1h-indazole-5-carboxylic acid ethyl ester, 1h-indazole-5-carboxylic acid, ethyl ester, 5-ethoxycarbonyl-1h-indazole, 1h-indazole-5-carboxylicacidethylester, pubchem18041, 5-ethoxycarbonylindazole, acmc-1c4yw, ethyl indazole-5-carboxylate, ethyl 2h-indazole-5-carboxylate, indazole-5-carboxylic acid ethyl ester
PubChem CID 1501980
IUPAC Name ethyl 1H-indazole-5-carboxylate
SMILES CCOC(=O)C1=CC=C2NN=CC2=C1

Specifications

Melting Point 122°C to 124°C
Quantity 1 g
Formula Weight 190.2
Percent Purity 95%
Chemical Name or Material Ethyl 1H-indazole-5-carboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.