missing translation for 'onlineSavingsMsg'
Learn More
Ethyl 2-chloronicotinate, 98%, Thermo Scientific Chemicals Product Code.: 11474257

Ethyl 2-chloronicotinate, 98%, Thermo Scientific Chemicals

Product Code. 11474257
10 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11474257

Brand: Thermo Scientific Alfa Aesar B20359.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ethyl 2-chloronicotinate,is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2-chloronicotinate,is used as a pharmaceutical intermediate.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1452-94-4
Molecular Formula C8H8ClNO2
Molecular Weight (g/mol) 185.607
MDL Number MFCD00674129
InChI Key PMIMPBYTPPRBGD-UHFFFAOYSA-N
Synonym ethyl 2-chloronicotinate, 2-chloronicotinic acid ethyl ester, ethyl-2-chloronicotinate, 2-chloro-nicotinic acid ethyl ester, ethyl 2-chloro-3-pyridinecarboxylate, 3-pyridinecarboxylic acid, 2-chloro-, ethyl ester, 2-chloro-3-pyridinecarboxylic acid ethyl ester, ethyl 2chloronicotinate, acmc-1brbe, ethyl 2-chloro-nicotinateShow MoreShow Less
PubChem CID 2796614
IUPAC Name ethyl 2-chloropyridine-3-carboxylate
SMILES CCOC(=O)C1=C(N=CC=C1)Cl

Specifications

Quantity 10 g
Formula Weight 185.61
Percent Purity 98%
Chemical Name or Material Ethyl 2-chloronicotinate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10247265 Certificate TypeCertificate of Analysis Date07/07/2023 Catalog NumberB20359,B20359.09,B20359.18
Lot Number5001V23X Certificate TypeCertificate of Analysis Date24/04/2023 Catalog NumberB20359.09,B20359.18
Lot Number10102405 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB20359.09,B20359.18
Lot NumberH2258A Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB20359.09,B20359.18
Lot Number10097604 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB20359.09,B20359.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl 2-chloronicotinate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.