missing translation for 'onlineSavingsMsg'
Learn More
Ethyl 2-methyl-4-pentenoate, 98%, Thermo Scientific Chemicals Product Code.: 11434097

Ethyl 2-methyl-4-pentenoate, 98%, Thermo Scientific Chemicals

Product Code. 11434097
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11434097

Brand: Thermo Scientific Alfa Aesar A19526.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53399-81-8
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD00010235
InChI Key BDBGKYIBDXAVMX-UHFFFAOYNA-N
Synonym ethyl 2-methyl-4-pentenoate, 2-methyl-4-pentenoic acid ethyl ester, ethyl 2-methyl 4-pentenoate, 4-pentenoic acid, 2-methyl-, ethyl ester, ethyl 2-methylpent-4-en-1-oate, ethyl 2-methyl-4-pentanoate, fema no. 3489, pineapple pentenoate, acmc-1ao14, ethyl-2-methyl-4-pentenoateShow MoreShow Less
PubChem CID 62024
IUPAC Name ethyl 2-methylpent-4-enoate
SMILES CCOC(=O)C(C)CC=C

Specifications

Density 0.88
Boiling Point 153°C to 155°C
Flash Point 40°C (104°F)
Refractive Index 1.416
Quantity 25 g
UN Number UN3272
Beilstein 1753406
Formula Weight 142.2
Percent Purity 98%
Chemical Name or Material Ethyl 2-methyl-4-pentenoate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10153534 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA19526.14,A19526.22
Lot Number90000355 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA19526.14,A19526.22
Lot Number10178743 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA19526.14,A19526.22
Lot Number10227038 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA19526.14,A19526.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl 2-methyl-4-pentenoate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.