Learn More
Ethyl 3-hydroxybenzoate, 99%, Thermo Scientific Chemicals Product Code.: 11468256
Ethyl 3-hydroxybenzoate, 99%, Thermo Scientific Chemicals
Product Code. 11468256
100 g
Click to view available options
Quantity:
25 g
100 g
500 g
Product Code. 11468256
Brand: Thermo Scientific Alfa Aesar A15343.22
Request Bulk or Custom Format
CAS: 7781-98-8 | C9H10O3 | 166.176 g/mol
Ethyl 3-hydroxybenzoate has been used in the synthesis of chiral derivatives of ethyl 3-(2-methylbutyloxy)benzoate and 4-(2-methylbutyloxy)-4'-acetylbiphenyl, via Mitsunobu reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethyl 3-hydroxybenzoate has been used in the synthesis of chiral derivatives of ethyl 3-(2-methylbutyloxy)benzoate and 4-(2-methylbutyloxy)-4′-acetylbiphenyl, via Mitsunobu reaction. It is a food-grade antimicrobial agent.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7781-98-8
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00002296
InChI Key MWSMNBYIEBRXAL-UHFFFAOYSA-N
Synonym ethyl-3-hydroxybenzoate, benzoic acid, 3-hydroxy-, ethyl ester, 3-hydroxybenzoic acid ethyl ester, m-ethoxycarbonylphenol, ethyl m-hydroxybenzoate, 3-hydroxy-benzoic acid ethyl ester, benzoic acid, m-hydroxy-, ethyl ester, pubchem19923, acmc-209pbw, ethyl 3-oxidanylbenzoate
PubChem CID 24522
IUPAC Name ethyl 3-hydroxybenzoate
SMILES CCOC(=O)C1=CC(=CC=C1)O
Specifications
Melting Point 72°C to 76°C
Boiling Point 187°C to 188°C (31 mmHg)
Quantity 100 g
Beilstein 2208566
Solubility Information Slightly soluble in water.
Formula Weight 166.18
Percent Purity 99%
Chemical Name or Material Ethyl 3-hydroxybenzoate
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10251542 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA15343.14,A15343.36,A15343.22
Lot Number10245410 Certificate TypeCertificate of Analysis Date18/01/2023 Catalog NumberA15343.14,A15343.36,A15343.22
Lot Number10240645 Certificate TypeCertificate of Analysis Date21/05/2022 Catalog NumberA15343.14,A15343.36,A15343.22
Lot Number10212397 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15343.14,A15343.36,A15343.22
Lot Number10158901 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15343.14,A15343.36,A15343.22
Product Title
Ethyl 3-hydroxybenzoate, 99%, Thermo Scientific Chemicals >