missing translation for 'onlineSavingsMsg'
Learn More
Ethyl 3-pyridineacetate, 99%, Thermo Scientific Chemicals Product Code.: 11437836

Ethyl 3-pyridineacetate, 99%, Thermo Scientific Chemicals

Product Code. 11437836
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11437836

Brand: Thermo Scientific Alfa Aesar A14768.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39931-77-6
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
MDL Number MFCD00006411
InChI Key RPWXYCRIAGBAGY-UHFFFAOYSA-N
Synonym ethyl 3-pyridylacetate, ethyl 2-pyridin-3-yl acetate, ethyl 3-pyridineacetate, 3-pyridineacetic acid, ethyl ester, acetic acid, 2-3-pyridyl-, ethyl ester, pyridin-3-yl-acetic acid ethyl ester, 3-pyridylacetic acid ethyl ester, ethyl 2-3-pyridyl acetate, pubchem20506, ethyl-3-pyridyl-acetateShow MoreShow Less
PubChem CID 96490
IUPAC Name ethyl 2-pyridin-3-ylacetate
SMILES CCOC(=O)CC1=CN=CC=C1

Specifications

Density 1.086
Boiling Point 79°C to 80°C (0.4 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5
Quantity 5 g
Beilstein 123827
Formula Weight 165.19
Percent Purity 99%
Chemical Name or Material Ethyl 3-pyridineacetate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0403995 Certificate TypeCertificate of Analysis Date08/04/2024 Catalog NumberA14768.06,A14768.14
Lot Number10213009 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14768.06,A14768.14
Lot Number10185900 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14768.06,A14768.14
Lot Number10138357 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14768.06,A14768.14
Lot Number10185902 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14768.06,A14768.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl 3-pyridineacetate, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.