missing translation for 'onlineSavingsMsg'
Learn More
Ethyl 7-methoxybenzo[b]furan-2-carboxylate, 97% Product Code.: 11400167

Ethyl 7-methoxybenzo[b]furan-2-carboxylate, 97%

Product Code. 11400167
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11400167

Brand: Thermo Scientific Alfa Aesar B21066.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 50551-58-1 | C12H12O4 | 220.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50551-58-1
Molecular Formula C12H12O4
Molecular Weight (g/mol) 220.22
MDL Number MFCD00060510
InChI Key SXCMZYVXJUJIDJ-UHFFFAOYSA-N
PubChem CID 521284
IUPAC Name ethyl 7-methoxy-1-benzofuran-2-carboxylate
SMILES CCOC(=O)C1=CC2=CC=CC(OC)=C2O1

Specifications

Melting Point 83°C to 86°C
Quantity 5 g
Synonym ethyl 7-methoxybenzofuran-2-carboxylate, 7-methoxybenzofuran-2-carboxylic acid ethyl ester, 7-methoxybenzofuran-2-carboxylic acid, ethyl ester, ethyl 7-methoxybenzo b furan-2-carboxylate, 7-methoxybenzofuran-2-carboxylic acidethyl ester, ethyl7-methoxybenzofuran-2-carboxylate, acmc-1aowz, 2-ethoxycarbonyl-7-methoxybenzofuran, ethyl 4-methoxylbenzofunan-2-carboxylate, ethyl 7-methoxybenzo d furan-2-carboxylate
Formula Weight 220.22
Percent Purity 97%
Chemical Name or Material Ethyl 7-methoxybenzo[b]furan-2-carboxylate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10132332 Certificate TypeCertificate of Analysis Date22/11/2023 Catalog NumberB21066.03,B21066.14,B21066.06
Lot Number10132332 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21066.03,B21066.14,B21066.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl 7-methoxybenzo[b]furan-2-carboxylate, 97% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.