Learn More
Ethyl (S)-(-)-1-Boc-4-oxopiperidine-2-carboxylate, 95%, Thermo Scientific Chemicals
70.55€ - 226.00€
Chemical Identifiers
CAS | 180854-44-8 |
---|---|
Molecular Formula | C13H21NO5 |
Molecular Weight (g/mol) | 271.313 |
MDL Number | MFCD15071881 |
InChI Key | YAVQLRUBKDCOCI-JTQLQIEISA-N |
Synonym | ethyl s-1-boc-4-oxopiperidine-2-carboxylate, s-1-tert-butyl 2-ethyl 4-oxopiperidine-1,2-dicarboxylate, 1-tert-butyl 2-ethyl 2s-4-oxopiperidine-1,2-dicarboxylate, ethyl r-+-1-boc-4-oxopiperidine-2-carboxylate, ethyl s---1-boc-4-oxopiperidine-2-carboxylate, ethyl s-n-tert-butoxycarbonyl-4-oxo-2-pipecolate, o1-tert-butyl o2-ethyl 2s-4-oxopiperidine-1,2-dicarboxylate, 2s-4-oxopiperidine-1,2-dicarboxylic acid 1-tert-butyl 2-ethyl ester |
PubChem CID | 11369276 |
IUPAC Name | 1-O-tert-butyl 2-O-ethyl (2S)-4-oxopiperidine-1,2-dicarboxylate |
SMILES | CCOC(=O)C1CC(=O)CCN1C(=O)OC(C)(C)C |
Description
1-Boc-4-oxopiperidine-2-carboxylate participates in the one-pot in situ formation and reaction of trimethyl (trichloromethyl) silane which finds application in the synthesis of 2, 2, 2-trichloromethylcarbinols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Boc-4-oxopiperidine-2-carboxylate participates in the one-pot in situ formation and reaction of trimethyl (trichloromethyl) silane which finds application in the synthesis of 2, 2, 2-trichloromethylcarbinols.
Notes
Store in a cool ventilated place. Avoid strong oxidizing materials.
Chemical Identifiers
180854-44-8 | |
271.313 | |
YAVQLRUBKDCOCI-JTQLQIEISA-N | |
11369276 | |
CCOC(=O)C1CC(=O)CCN1C(=O)OC(C)(C)C |
C13H21NO5 | |
MFCD15071881 | |
ethyl s-1-boc-4-oxopiperidine-2-carboxylate, s-1-tert-butyl 2-ethyl 4-oxopiperidine-1,2-dicarboxylate, 1-tert-butyl 2-ethyl 2s-4-oxopiperidine-1,2-dicarboxylate, ethyl r-+-1-boc-4-oxopiperidine-2-carboxylate, ethyl s---1-boc-4-oxopiperidine-2-carboxylate, ethyl s-n-tert-butoxycarbonyl-4-oxo-2-pipecolate, o1-tert-butyl o2-ethyl 2s-4-oxopiperidine-1,2-dicarboxylate, 2s-4-oxopiperidine-1,2-dicarboxylic acid 1-tert-butyl 2-ethyl ester | |
1-O-tert-butyl 2-O-ethyl (2S)-4-oxopiperidine-1,2-dicarboxylate |
Specifications
180854-44-8 | |
>110°C (230°F) | |
1.46 | |
ethyl s-1-boc-4-oxopiperidine-2-carboxylate, s-1-tert-butyl 2-ethyl 4-oxopiperidine-1,2-dicarboxylate, 1-tert-butyl 2-ethyl 2s-4-oxopiperidine-1,2-dicarboxylate, ethyl r-+-1-boc-4-oxopiperidine-2-carboxylate, ethyl s---1-boc-4-oxopiperidine-2-carboxylate, ethyl s-n-tert-butoxycarbonyl-4-oxo-2-pipecolate, o1-tert-butyl o2-ethyl 2s-4-oxopiperidine-1,2-dicarboxylate, 2s-4-oxopiperidine-1,2-dicarboxylic acid 1-tert-butyl 2-ethyl ester | |
−20° (c=0.5 in Chloroform) | |
1-O-tert-butyl 2-O-ethyl (2S)-4-oxopiperidine-1,2-dicarboxylate | |
11369276 | |
95% |
1.079 | |
C13H21NO5 | |
MFCD15071881 | |
YAVQLRUBKDCOCI-JTQLQIEISA-N | |
CCOC(=O)C1CC(=O)CCN1C(=O)OC(C)(C)C | |
271.313 | |
271.31 | |
Ethyl (S)-(-)-1-Boc-4-oxopiperidine-2-carboxylate |
Safety and Handling
GHS H Statement
H319
Causes serious eye irritation.
P264b-P280i-P305+P351+P338
H319
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only