missing translation for 'onlineSavingsMsg'
Learn More

Ethylbenzene, puriss. p.a., ≥99.0% (GC), Honeywell™ Riedel-de-Haën™

51.46€ - 132.40€

Chemical Identifiers

CAS 100-41-4
Molecular Formula C8H10
Molecular Weight (g/mol) 106.168
MDL Number MFCD00011647
InChI Key YNQLUTRBYVCPMQ-UHFFFAOYSA-N
Synonym phenylethane, ethylbenzol, benzene, ethyl, aethylbenzol, ethylenzene, ethyl benzene, ethylbenzeen, etilbenzene, etylobenzen, alpha-methyltoluene
PubChem CID 7500
ChEBI CHEBI:16101
IUPAC Name ethylbenzene
SMILES CCC1=CC=CC=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15650660
View Documents
Honeywell Riedel-de Haën
03080-250ML
250 mL
51.46€
250mL
In Stock
to see stock.
Add to basket
 
15640660
View Documents
Honeywell Riedel-de Haën
03080-1L
1 L
132.40€
1 L
In Stock
to see stock.
Add to basket
 
Description

Description

Specifications

Chemical Identifiers

100-41-4
106.168
YNQLUTRBYVCPMQ-UHFFFAOYSA-N
7500
ethylbenzene
C8H10
MFCD00011647
phenylethane, ethylbenzol, benzene, ethyl, aethylbenzol, ethylenzene, ethyl benzene, ethylbenzeen, etilbenzene, etylobenzen, alpha-methyltoluene
CHEBI:16101
CCC1=CC=CC=C1

Specifications

100-41-4
15°C (59°F)
C6H5C2H5
UN1175
phenylethane, ethylbenzol, benzene, ethyl, aethylbenzol, ethylenzene, ethyl benzene, ethylbenzeen, etilbenzene, etylobenzen, alpha-methyltoluene
CCC1=CC=CC=C1
106.168
CHEBI:16101
≥99.0% (GC)
Ethylbenzene
0.867g/mL at 25°C
C8H10
MFCD00011647
1901871
YNQLUTRBYVCPMQ-UHFFFAOYSA-N
ethylbenzene
7500
106.17g/mol
puriss. p.a.
Videos
Safety and Handling

Safety and Handling

P210-P261-P273-P301 + P310-P304 + P340 + P312-P331

H225-H304-H332-H373-H412

missing translation for 'dotInformation' : class 3 - PG 2 - Ethylbenzene

missing translation for 'einecsNumber' : 202-849-4

missing translation for 'rtecsNumber' : DA0700000

Product Identifier
  • Ethylbenzene

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H332-Harmful if inhaled.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : ethylbenzene

SDS
Documents

Documents

Product Certifications