Learn More
IKA™ EUROSTAR 100 Control P4
GREENER_CHOICE
EUROSTAR 100 Control P4 is a powerful electronic overhead stirrer.
Brand:  IKA™ 0025004384
Product Code. 16258641
  • 5185.00€ / Each
Estimated Shipment: 30-01-2025
Description
  • Reverse function
  • Multilingual TFT display
  • Programmable functions
  • Integrated temperature measurement
  • Interval operation
  • Timer function
  • Adjustable safety circuit
  • Locked function
  • Infinitely adjustable speed
  • Push-through agitator shafts
  • Overload protection
  • Short-term overload operation
  • Quiet operation
  • Error code Display
  • H 67.60 temperature sensor and WH 11 WiCo holder included in delivery
Specifications
Specifications
10 to 300 rpm
TFT
174 w
Brushless DC
Max. 320 Ncm
100 L
100/230 V
86 x 352 x 230 mm
-10°C to +350°C
Max. 100,000 mPas