Learn More
Thermo Scientific™ Eutech™ pH 700 Meter
Economical, user-friendly and accurate, the Eutech pH 700 is your ideal choice for routine applications in laboratories, productions plants and schools.
Brand:  Thermo Scientific™ 01X541603
 View more versions of this product
Product Code. 16385985
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Economical, user-friendly and accurate, the Eutech pH 700 is your ideal choice for routine applications in laboratories, productions plants and schools. It features a large, easy-to-read display, ready indicator that letsyou know when readings are stable and up to 5-point calibration with auto-buffer recognition. It also has a non-volatile memory holds up to 100 data points and comes witu an integral electrode holder.
Applications:
  • Educational: Useful for most laboratory, ecological studies and other applications.
  • Laboratory: Environmental studies, chemical labs, titrations and quality assurance testing. Use in all types of food processing.
Buffer Sets
  • USA (1.68, 4.01, 7.00, 10.01, 12.45)
  • NIST (1.68, 4.01, 6.86, 9.18, 12.45)
  • TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    Eutech pH 700 Meter With Integral Electrode Holder & 100/240 VAC Adapter
    ±0.01
    0.01
    Up to 5
    32.0°F to 212.0°F
    Offset in 0.1 °increments; offset range: ±5.0 °C / ±9.0 °F
    ORP: ±0.2 mV (±199.9 mV) / ±2 mV (beyond)
    ±2000 mV
    ATC/MTC (0°C to 100°C) (pH only)
    ±0.3°C/±0.5°F
    -2 to 16
    ±2000 mV
    0°C to 100°C
    0.1°C/°F
    9 V DC adapter, 1.3 A
    100/240 VAC, SMPS
    ORP: 0.1 mV (±199.9 mV) / 1 mV (beyond)