Learn More
Novus Biologicals™ EV MaxiSEC Columns
920.00€
Specifications
Content And Storage | Store at 4°C. |
---|---|
Sample Type | Urine, cell culture supernatant, or other diluted matrices |
Description
Specifications
Store at 4°C. | |
Urine, cell culture supernatant, or other diluted matrices |
Product Title
Novus Biologicals™ EV MaxiSEC Columns