Learn More
Novus Biologicals™ EV SEC Columns
529.00€ - 920.00€
Specifications
Content And Storage Store at 4°C.
Sample Type Biofluids or cell culture supernatant
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18620311
View Documents
Novus Biologicals™
NBP3-11763-10Units
10 Columns
920.00€
Pack of 10
18694320
View Documents
Novus Biologicals™
NBP3-11763-5Units
5 Columns
529.00€
Pack of 5
Specifications
Store at 4°C.
Biofluids or cell culture supernatant
Documents
Product Certifications
Product Content Correction
Product Title
Novus Biologicals™ EV SEC Columns