Learn More
Novus Biologicals™ Exosome Standards (K-562 cell line)
Highly pure, lyophilized exosome standards with superior stability, optimal for multiple applications
Brand:  Novus Biologicals™ NBP2-49864-60ug
 View more versions of this product
Product Code. 18624050
  • 417.00€ / Pack of 2
Estimated Shipment: 08-01-2025
Specifications
Store at 4°C. Do not freeze.
LYOPH
2 x 30 μg Vials