Learn More
Novus Biologicals™ Exosome Standards (NCI-H1975 cell line)
Highly pure, lyophilized exosome standards with superior stability, optimal for multiple applications
Brand:  Novus Biologicals™ NBP3-11688-180ug
 View more versions of this product
Product Code. 18642921
  • 1080.00€ / Pack of 6
Estimated Shipment: 03-02-2025
Specifications
Store at 4°C. After reconstitution store at -70°C.
LYOPH
6 x 30 μg Vials