Learn More
Taylor Wharton™ XT Series Long-Term Cryogenic Storage Dewar
The durable ribbed high-strength aluminium body, and superior vacuum performance and insulation provide versatile extended-time storage.
1490.00€ - 1780.00€
Specifications
Material | Aluminum |
---|---|
Description | Long-term Cyrogenice Storage Dewar |
For Use With (Application) | Cryogenic storage |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
11768509
|
Taylor Wharton™
10XTB-11M |
10 L |
1610.00€
Each |
In Stock
Log in to see stock. ![]() |
||||||
11778509
|
Taylor Wharton™
XT2011 |
20 L |
1670.00€
Each |
Estimated Shipment: 15-09-2025
Log in to see stock. |
||||||
11788509
|
Taylor Wharton™
34XTB11 |
34 L |
1780.00€
Each |
Estimated Shipment: 01-09-2025
Log in to see stock. |
||||||
11758509
|
Taylor Wharton™
XTL8 |
8 L |
1490.00€
Each |
Estimated Shipment: 15-09-2025
Log in to see stock. |
||||||
Description
- Provides extended-time storage
- High-strength aluminium body
- Provides maximum holding times with superior vacuum performnce and insulation
Specifications
Aluminum | |
Long-term Cyrogenice Storage Dewar | |
Cryogenic storage |
Product Title
Taylor Wharton™ XT Series Long-Term Cryogenic Storage Dewar