Learn More
Lenz Laborglasintrumente Extractors Acc. to Knöfler-Böhm
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 05375043
 View more versions of this product
Product Code. 10204011
  • 461.00€ / Each
Only null left
Estimated Shipment: 17-09-2025
Description
  • The complete apparatuses are consisting of: Jacketed extractor head made of DURAN™ tubing, Dimroth condenser (details on page 71)
  • With thread GL 14
  • Made of DURAN™ tubing
  • Round bottom flask
Specifications
Specifications
150 mL
Duran Glass
NS 45/40
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Lenz Laborglasintrumente Extractors Acc. to Knöfler-Böhm > 150 mL