Learn More
Esculape Laboratoire™ Eyewash Station
Administer emergency eye care using an Esculape™ Eyewash Station. Components of this station include those that rinse and evacuate foreign substances from the eyes.
63.00€ - 72.17€
Specifications
For Use With (Application) | For Remove Particles or Dust, Rehydrates the Eye |
---|---|
Product Type | Eyewash Station |
Product Code | Brand | Includes | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | ||||||
10617204
|
Esculape Laboratoire™
8205201 |
0.9% Sodium Chloride, 2 x 500 mL | N/A | |||||||
11769522
|
Esculape Laboratoire™
8205200 |
0.9% Sodium Chloride, 500 mL |
63.00€
Each |
In Stock
Log in to see stock. ![]() |
||||||
Description
- Fixed position with wall bracket;
- Ergonimic eye cup aids continuous rinsing
- Quick release bottle caps
- Instructions in three languages
- Three year shelf life if unopened
Specifications
For Remove Particles or Dust, Rehydrates the Eye | |
Eyewash Station |
Product Title
Esculape Laboratoire™ Eyewash Station