Learn More
Thermo Scientific™ EZ-Link™ Biocytin
Synthesize long or trifunctional biotinylation reagents using this biotin variant containing a primary amine in its valeric acid chain.
Brand: Thermo Scientific™ 28022
Description
Thermo Scientific™ EZ-Link Biocytin is a simple variant of biotin that contains a primary amine in its valeric acid chain, which provides the essential backbone for construction of certain long-chain and trifunctional biotinylation reagents.
Highlights:
- Useful for synthesizing long-chain biotinylation reagents (can use EDC, Part No. 22980, 22981)
- An extended spacer arm intermediate for further derivatization into reactive group-specific biotinylating reagents
- Contains both carboxyl group and primary amine, which may be modified to make trifunctional reagents like Sulfo-SBED

Specifications
Biocytin | |
Store at room temperature. | |
Cell-Impermeant | |
Water | |
Mid-length |
100 mg | |
EZ-Link™ | |
Amine, Carboxylic Acid | |
Biotin Analog | |
Biotin & Analogs |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.