missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EZ-Link™ Biotin-LC-Hydrazide

Biotinylate glycoproteins (aldehydes of oxidized sugars) with this mid-length (24.7A) hydrazide-activated biotinylation reagent.

Brand:  Thermo Scientific™ 21340

Product Code. 11881205

  • 315.00€ / 50mg
Estimated Shipment: 17-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ EZ-Link Hydrazide-LC-Biotin is a mid-length, simple, hydrazide-activated biotinylation reagent for labeling glycoproteins and other carbohydrate-containing compounds having oxidizable sugars or aldehydes.

Biotin hydrazide labeling reagents enable simple and efficient biotin labeling of antibodies and other glycoproteins.

Highlights:

  • Carbohydrate-reactive
  • Labels immunoglobulins in the Fc region
  • Long spacer arm helps prevent steric hindrances
  • Must be dissolved in DMSO (50mM) before adding to aqueous buffer
  • Use pH 4-6 to complete coupling reaction
  • Forms hydrazone bond
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Biotin-LC-Hydrazide
Store at 4°C.
Cell-Permeant
Amine, Hydrazide
Biotin
Biotin & Analogs
50 mg
EZ-Link™
Carboxylic Acid, Ketone, Aldehyde
DMF (Dimethylformamide), DMSO (Dimethylsulfoxide)
Mid-length
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.