Learn More
Thermo Scientific™ EZ-Link™ TFP-PEG3-Biotin
Biotinylate primary amines (e.g., protein lysines) and secondary amines such as histidine and tryptophan with this tetrafluorophenyl-ester biotinylation reagent.
Brand: Thermo Scientific™ 21219
This item is not returnable.
View return policy
Description
Thermo Scientific™ EZ-Link TFP-PEG3-Biotin is a tetrafluorophenyl-ester biotinylatoin reagent that enables labeling at primary amines found on lysine residues and the N-terminus and secondary amines such as histidine and tryptophan.
Highlights:
- Tetrafluorophenyl (TFP) ester reacts with primary and secondary amines at pH 7-9
- Long-chain, water-soluble, polyethylene glycol (PEG) spacer arm
- PEG spacer arm transfers hydrophilicity to final conjugate
- Creates an amide bond
Specifications
No | |
Tetrafluorophenyl ester of biotin with a triethyleneglycol spacer | |
No | |
32.6 | |
Approx. 25 uses to label a total of 480mg of IgG using typical conditions | |
Water |
EZ-Link TFP-PEG3-Biotin | |
694.74 | |
Yes | |
50 mg | |
Amine |