missing translation for 'onlineSavingsMsg'
Learn More
Learn More
F4/80, Rabbit anti-Mouse, Clone: Cl:A3-1 (recombinant version), Novus Biologicals™
Rabbit Monoclonal Antibody
261.00€ - 569.00€
Specifications
Antigen | F4/80 |
---|---|
Clone | Cl:A3-1 (recombinant version) |
Concentration | 1 mg/mL |
Dilution | Flow Cytometry, Immunohistochemistry |
Applications | Flow Cytometry, Immunohistochemistry |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18392501
|
Bio-Techne
NBP2-81029-0.025MG |
0.025 mg |
261.00€
25µg |
Estimated Shipment: 30-01-2025
Log in to see stock. |
|||||
18236269
|
Bio-Techne
NBP2-81029-0.2MG |
0.2 mg |
569.00€
0.20mg |
Estimated Shipment: 30-01-2025
Log in to see stock. |
|||||
Description
F4/80 Monoclonal specifically detects F4/80 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry.Specifications
F4/80 | |
1 mg/mL | |
Flow Cytometry, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Adaptive Immunity, GPCR, Immune System Diseases, Immunology, Innate Immunity, Myeloid derived Suppressor Cell | |
PBS | |
13733 | |
Thioglycollate stimulated peritoneal macrophages of mouse origin. | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Cl:A3-1 (recombinant version) | |
Flow Cytometry, Immunohistochemistry | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
ADGRE1, cell surface glycoprotein F4/80, DD7A5-7, EGF-like module containing, mucin-like, hormone receptor-like sequence 1, EGF-like module receptor 1, EGF-like module-containing mucin-like hormone receptor-like 1, EGF-TM7, EMR1 hormone receptor, F4/80, Gpf480, Ly71, lymphocyte antigen 71, Mouse EMR1, Rat EMR1, TM7LN3 | |
Adgre1 | |
IgG κ | |
Protein A purified | |
This antibody recognises the mouse F4/80 antigen, a 160kD glycoprotein, expressed by murine macrophages. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title