missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM151B Monoclonal Antibody (OTI2B3), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA527476
This item is not returnable.
View return policy
Description
FAM151B is a protein coding gene.Specifications
FAM151B | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6UXP7 | |
FAM151B | |
Full length human recombinant protein of FAM151B produced in HEK293T cell | |
100μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI2B3 | |
Unconjugated | |
FAM151B | |
4930405M20Rik; AASA9217; f151; FAM151B; family with sequence similarity 151 member B; family with sequence similarity 151, member B; protein FAM151B; UNQ9217; UNQ9217/PRO28685 | |
Mouse | |
Affinity Chromatography | |
RUO | |
167555 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction