missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Favipiravir Product Code.: 17291276
compliance-icons compliance-icons

Thermo Scientific Chemicals Favipiravir

Product Code. 17291276
100 mg
Click to view available options
Quantity:
100 mg
500 mg
This item is not returnable. View return policy

Product Code. 17291276

Brand: Thermo Scientific Chemicals 468691000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 259793-96-9 | C5H4FN3O2, C5H4FN3O2 | 157.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 259793-96-9
Molecular Formula C5H4FN3O2

Specifications

Quantity 100 mg
Boiling Point 52.6°C
Formula Weight 157.09
InChI Key ZCGNOVWYSGBHAU-UHFFFAOYSA-N
SMILES NC(=O)C1=NC(F)=CNC1=O
IUPAC Name 6-fluoro-3-oxo-3,4-dihydropyrazine-2-carboxamide
Molecular Weight (g/mol) 157.10
Chemical Name or Material Favipiravir
compliance-icons compliance-icons
Product Identifier
  • Favipiravir
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H228-Flammable solid.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
Supplemental information
  • MIXTURE LIST-Contains : Favipiravir

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Favipiravir >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.