Learn More
Thermo Scientific™ NoLimits 3000 bp DNA Fragment
GREENER_CHOICE
Optimize applications where a custom DNA size standard is required with NoLimits, a collection of 36 chromatography-purified individual DNA fragments ranging from 10 -20,000bp.
Brand:  Thermo Scientific™ SM1711
60.97 EUR valid until 2025-06-30
Use promo code "24108" to get your promotional price.
Product Code. 10766411
  • 91.00€ / 10µg
Nurture Your Progress
Use promo code 24108 at checkout
View Available Promotions
Description
NoLimits is a collection of 36 highly-purified individual DNA fragments ranging from 10 to 20,000 bp. Unlike other vendors, where custom standards are produced by PCR, Thermo Scientific NoLimits DNA fragments are produced using specifically designed plasmids purified by patented technology.
Plasmid DNA is digested with Thermo Scientific Restriction Enzymes, and the individual DNA fragments are chromatography-purified from the digestion mixture. As a result, we offer exceptionally pure DNA fragments that are free of any truncated or degraded molecules. Our custom DNA fragments are the best choice for applications, ranging from electrophoresis (both gel and capillary) to HPLC and beyond.
Highlights
  • Chromatography-purified individual DNA fragments
  • Broad range: 10 to 20,000 bp
  • Sharp peaks during capillary electrophoresis, sharp bands during gel electrophoresis
  • Precise DNA concentration 
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Agarose
No
Agarose Gels
NoLimits
DNA Fragment
10 to 20000 bp
Sustainable packaging
10 μg
SDS
Documents Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2839140 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberSM1711
Lot Number2924098 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog NumberSM1711
Lot Number3029205 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog NumberSM1711
Lot Number3085551 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog NumberSM1711
Lot Number3085558 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog NumberSM1711
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ NoLimits 3000 bp DNA Fragment > 10μg