Learn More
Tocris Bioscience™ FFN 102 mesylate
Selective fluorescent substrate of DAT and VMAT2
Brand:  Tocris Bioscience™ 5200/10
214.25 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18788788
  • 272.00€ / 10mg
Estimated Shipment: 12-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
FFN 102 mesylate is a pH responsive fluorescent false neurotransmitter (FFN). Selective dopamine transporter (DAT) and VMAT2 substrate. Exhibits no significant binding to a panel of 38 CNS receptors, including dopamine and serotonin receptors. Inhibits dopamine uptake. Excitation maxima are 340 nm at pH 5 and 370 nm at pH 7.5. Emission maximum is 435 nm at both pH 5 and 7.5.
Specifications
Specifications
>99%
10 mg
335.76
1883548-92-2
FFN 102 mesylate
FFN 102 mesylate