missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF-10 Antibody (014), HRP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89572H
This item is not returnable.
View return policy
Description
FGF-10 Monoclonal antibody specifically detects FGF-10 in Human samples. It is validated for ELISA
Specifications
FGF-10 | |
Monoclonal | |
HRP | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
014 | |
ELISA | |
FGF-10, fibroblast growth factor 10, Keratinocyte growth factor 2, produced by fibroblasts of urinary bladder lamina propria | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human FGF-10 (NP_004456.1; Gln38-Ser208). | |
0.1 mL | |
Angiogenesis, Biologically Active Proteins, Cell Cycle and Replication | |
2255 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
FGF-10 Antibody (014), HRP, Novus Biologicals™ > 0.1 mL; HRP
Spot an opportunity for improvement?Share a Content Correction