missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF basic/FGF2/bFGF Antibody (S03-9C3), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20011-50ul
This item is not returnable.
View return policy
Description
FGF basic/FGF2/bFGF Monoclonal antibody specifically detects FGF basic/FGF2/bFGF in Human samples. It is validated for Western Blot
Specifications
FGF basic/FGF2/bFGF | |
Monoclonal | |
0.3 mg/mL | |
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
S03-9C3 | |
Unconjugated | |
Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2 | |
Recombinant protein of human FGF basic/FGF2/bFGF (Uniprot # P09038) | |
50 μg | |
Angiogenesis, Biologically Active Proteins, Cancer | |
2247 | |
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
FGF basic/FGF2/bFGF Antibody (S03-9C3), Novus Biologicals™ > 50 μg; Unconjugated
Spot an opportunity for improvement?Share a Content Correction