Learn More
Thermo Scientific Chemicals Fibronectin CS1 Peptide
For cellular and molecular biology applications
Brand: Thermo Scientific Chemicals J66606.M^
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
HPLC | |
Solid | |
Fibronectin CS1 Peptide | |
RUO - research use only |
-30°C to -10°C | |
872.96 | |
1 mg | |
Glu-Ile-Leu-Asp-Val-Pro-Ser-Thr |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
X18K034 | Certificate of Analysis | 05/11/2024 | J66606.#0,J66606.M^ |
X18K035 | Certificate of Analysis | 05/11/2024 | J66606.#0,J66606.M^ |
W26K013 | Certificate of Analysis | 07/10/2024 | J66606.#0,J66606.M^ |
P02K013 | Certificate of Analysis | 12/03/2024 | J66606.#0,J66606.M^ |
M25K003 | Certificate of Analysis | 05/02/2024 | J66606.#0,J66606.M^ |