missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Afora™ Filling Column
Used in general laboratory applications. Afora™ Filling Columns can be used to prepare samples for later use.
Brand: Afora™ 2517/E
This item is not returnable.
View return policy
Description
- For use with general laboratory applications, funneling, and filtration.
- Cylindrical shape
- One male ground glass joint at the top, one female ground glass joint at the bottom.
Specifications
40 mm | |
Glass | |
Ground glass joints: |
760 mL | |
Total: 735 mm, Working: 600 mm | |
NS 45/40 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Afora™ Filling Column >
Spot an opportunity for improvement?Share a Content Correction