Learn More
Honeywell™ FINEDEX™ 944-01 Clean Gloves
Click to view available options
Size: 8
Description
- Unlined yellow latex glove
- Cotton flock inside
- Diamond-pattern finish
- Natural latex provides excellent flexibility, synonymous with dexterity and comfort
- The cotton flocked interior provides the user with greater comfort
- Diamond finish is tailored for grasping dry and damp objects
- Latex offers good mechanical and chemical resistance against detergents and liquid products
- The cuff helps to protect a large part of the forearm
- The production process helps to obtain a glove with a minimum latex protein
Specifications
Specifications
Certifications/Compliance | EN 420-2003 EEC Basic standards, EN 388-2003 Mechanical hazards, EN 374-2003 Chemical hazards |
Color | Yellow |
Material | Latex, Cotton |
Size | 8 |
Thickness (Metric) | 0.4 to 0.8 mm |
Length (Metric) | 30 cm |