Learn More
Thermo Scientific™ Finger Tite Connectors
Ensure reliable, leak-free connections for optimal GC performance using these Finger Tite Connectors.
150.00€ - 377.00€
Specifications
For Use With (Equipment) TRACE GC Ultra and FOCUS GC Instruments
View More Specs
Products 3
Product Code Brand Type Price Quantity & Availability  
Product Code Brand Type Price Quantity & Availability  
10584465
View Documents
Thermo Scientific™
290ST130
Female Nut
150.00€
Pack of 5
Only null left
Estimated Shipment: 30-06-2025
Add to basket
10177804
View Documents
Thermo Scientific™
290ST131
Split/Splitless and MS Starter Kit This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A
10321485
View Documents
Thermo Scientific™
290ST132
Split/Splitless and FID Starter Kit This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A
Description
Description
Ensure reliable, leak-free connections for optimal GC performance using Thermo Scientific™ Finger Tite Connectors. The perfect metal-to-metal seal eliminates contamination and produces lower air/water background noise compared to conventional graphite/polymer ferrules. They are easy to handle even with a hot injector/detector and do not require the use of tools. Unlike other ferrules, they do not require retightening after installation.
Recommended Use
  • Thermo Scientific™ and Agilent™ GC instruments
  • Connectors for SSL and MS systems
Specifications
Specifications
TRACE GC Ultra and FOCUS GC Instruments
Documents
Product Certifications
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Product Content Correction
Product Title
Thermo Scientific™ Finger Tite Connectors