Learn More
Fisherbrand™ Replaceable Tip Probe for use with 500 and 700 watt Sonic Dismembrators
Microtips are thin, high intensity probes which are designed for processing small sample volumes. Microtips screw into the threaded end of the standard 1/2 in. probe (4220).
615.00€ - 662.00€
Specifications
Product Type | Probe |
---|---|
For Use With (Equipment) | Model 505/705 Sonic dismembrator |
Requires: Standard 1/2 in. probe (Cat. No. FB4220)
Product Code | Brand | Diameter (Metric) Tip | Volume Range (Metric) | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Tip | Volume Range (Metric) | Includes | Price | Quantity & Availability | ||||
12921171
|
Fisherbrand™
FB4220 |
13 mm | 20 to 250 mL | Standard 1/2 in. Probe |
615.00€
Each |
Estimated Shipment: 15-07-2025
Log in to see stock. |
||||
12971161
|
Fisherbrand™
FB4207 |
19 mm | 50 to 500 mL | Replacement tip |
631.00€
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||
12981161
|
Fisherbrand™
FB4208 |
19 mm | 50 to 500 mL | Probe |
631.00€
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||
12991161
|
Fisherbrand™
FB4209 |
25 mm | 100 mL to 1 L | Probe |
662.00€
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||
12901171
|
Fisherbrand™
FB4210 |
25 mm | 100 mL to 1 L | Replacement tip |
662.00€
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||
12911171
|
Fisherbrand™
FB4219 |
13 mm | 20 to 250 mL | Probe |
615.00€
Each |
In Stock
Log in to see stock. ![]() |
||||
Description
These Fisher Scientific™ Probes are designed for use with 500 and 700 watt Sonic Dismembrators.
- Available in diameters ranging from 1/16 in. to 1 in.
- Capacity options of 0.2-5mL to 100-1000mL
- Replaceable tip, solid and microtip probes
Specifications
Probe | |
Model 505/705 Sonic dismembrator |