Learn More
Fisherbrand™ 200 Series Safety Glasses
Shop All Fisherbrand Products
Click to view available options
Frame Color:
Black
Unit Size:
Pack of 12
Specifications
Specifications
Certifications/Compliance | ANSI Z87.1, ANSI Z80.3 |
Lens Tint | Clear |
Product Type | Safety Glass |
Frame Color | Black |
Temple Style | Adjustable; spatula |
Product Content Correction
Product Title
Fisherbrand™ 200 Series Safety Glasses >