Learn More
Fisherbrand™ Infecon™ 2000 and 2050 Infectious/Diagnostic Shippers
For shipping test tubes containing infectious substances or diagnostic specimens
160.00€
Specifications
Certifications/Compliance Complies with ICAO/IATA Packing Instruction 602
Applications For shipping test tubes containing infectious substances or diagnostic specimens
View More Specs
Products 1
Product Code Brand Type Size Price Quantity & Availability  
Product Code Brand Type Size Price Quantity & Availability  
15502066
View Documents
Fisherbrand™
19075384B
Infecon 2000, Infectious/Diagnostic Shipper 4 x 4 x 8 in.
160.00€
Pack of 6
Estimated Shipment: 30-06-2025
Description
For shipping test tubes containing infectious substances or diagnostic specimens
  • Bubble-wrap compartment
  • Liquid-tight, plastic secondary container withstands pressure up to 95kPa
  • U.N. markings and a label for both infectious substances and diagnostic specimens
Compliance
Complies with ICAO/IATA Packing Instruction 602
Specifications
Specifications
Complies with ICAO/IATA Packing Instruction 602
For shipping test tubes containing infectious substances or diagnostic specimens
Documents
Product Certifications
Product Content Correction
Product Title
Fisherbrand™ Infecon™ 2000 and 2050 Infectious/Diagnostic Shippers