Description
Value your lab can rely on
Spigots for use with Fisherbrand HDPE and Polypropylene rectangular carboys
- Spigots include a hose barb insert to accommodate -+ to 5/16-in. ID tubing
- Spigot spout without the adapter fits 5/8-in. ID tubing
Greener Choice Claims
- Environmental benefits include:
- Renewable Energy
Specifications
Specifications
Autoclavable | Autoclavable |
For Use With (Equipment) | Fisherbrand HDPE and Polypropylene Rectangular Carboys |
Material | Polypropylene |
No. per Case | 1 Ea. |
Product Type | Spigot |
Product Content Correction
Product Title
Fisherbrand™ Polypropylene, Spigot >